missing translation for 'onlineSavingsMsg'
Learn More

Dichlorofluorescein, 0.1% Alcoholic, Certified, LabChem™

Supplier:  LabChem LC135807

 View more versions of this product

Catalog No. LC135807


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dichlorofluorescein
54.3,39.89,3,2.7,0.1
C20H10Cl2O5
Passes Test
Certified
Passes Test
Yellow
Liquid
0.1% Alcoholic
7732-18-5 , 67-63-0 , 67-56-1 , 76-54-0
Soluble in water
401.2
Poly Bottle
Hydrogen fluoride; Carbon monoxide; Carbon dioxide; Hydrogen chloride
125 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions