missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichlorofluorescein, 0.1% Alcoholic, Certified, LabChem™
$105.14 - $215.04
Specifications
| Chemical Name or Material | Dichlorofluorescein |
|---|---|
| Name Note | 0.1% Alcoholic |
| CAS Max % | 54.3,39.89,3,2.7,0.1 |
| CAS | 7732-18-5,67-63-0,67-56-1,76-54-0 |
| Linear Formula | C20H10Cl2O5 |
Specifications
| Dichlorofluorescein | |
| 54.3,39.89,3,2.7,0.1 | |
| C20H10Cl2O5 | |
| Passes Test | |
| Certified | |
| Passes Test | |
| Yellow |
| 0.1% Alcoholic | |
| 7732-18-5,67-63-0,67-56-1,76-54-0 | |
| Soluble in water | |
| 401.2 | |
| Poly Bottle | |
| Hydrogen fluoride; Carbon monoxide; Carbon dioxide; Hydrogen chloride | |
| Liquid |