accessibility menu, dialog, popup

UserName

CAS RN 76-54-0

IUPAC Name: 2',7'-dichloro-3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: dichlorofluorescein
Molecular Weight (g/mol): 401.2
Molecular Formula: C20H10Cl2O5
InChi Key: VFNKZQNIXUFLBC-UHFFFAOYSA-N
SMILES: OC1=CC2=C(C=C1Cl)C1(OC(=O)C3=CC=CC=C13)C1=CC(Cl)=C(O)C=C1O2
Molecular Weight (g/mol) 
  • (11)
  • (5)
Quantity 
  • (3)
  • (3)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
Grade 
  • (6)
  • (2)
  • (3)
  • (1)
Color 
  • (4)
  • (4)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (4)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (7)
  • (11)
  • (5)
  • (3)
  • (3)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (6)
  • (2)
  • (3)
  • (1)
  • (4)
  • (4)
  • (1)
  • (2)
  • (2)