missing translation for 'onlineSavingsMsg'
Learn More

Dibromomethane 99.0+%, TCI America™

Catalog No. D0192500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0192500G 500 g
D019225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0192500G Supplier TCI America Supplier No. D0192500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 74-95-3
Molecular Formula CH2Br2
Molecular Weight (g/mol) 173.835
MDL Number MFCD00000168
InChI Key FJBFPHVGVWTDIP-UHFFFAOYSA-N
Synonym methylene bromide, methane, dibromo, methylene dibromide, dibromomethylene, rcra waste number u068, dibrommethan, methylenbromid, dibromo-methane, ccris 939, rcra waste no. u068
PubChem CID 3024
ChEBI CHEBI:47077
IUPAC Name dibromomethane
SMILES C(Br)Br

Specifications

Color Colorless
Boiling Point 97°C
Quantity 500 g
UN Number 2664
Formula Weight 173.84
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibromomethane
EINECSNumber (9)-1417
RTECSNumber PA7350000
TSCA Yes