missing translation for 'onlineSavingsMsg'
Learn More

Decon™ LeLoop™ Inoculating Loops
GREENER_CHOICE

More economical than platinum or disposable plastic loops

Supplier:  Decon™ MP19825

 Afficher plus de versions de ce produit

Numéro de catalogue. 22032091


Il en reste null

Description

Description

More economical than platinum or disposable plastic loops

Reusable loops are made of high-quality twisted nichrome wire.

  • Loops slide smoothly across agar surfaces without tearing or gouging
  • The nichrome wire cools rapidly, saving time
  • Loops are available calibrated or noncalibrated
  • Calibrated in accordance with ASM/APHA via Evans Blue Test and include calibration certificates
  • LeLoop™ loops come without holders, which are available separately
Spécifications

Identifiants chimiques

3 in.
Nichrome
Inoculating Loop
7.62 cm
Non-sterile

Spécifications

Non-sterile
7.62 cm
Nichrome
0.35 cm
Inoculating Loop
3 in.
5.45 μL
0.14 in.
Large
No
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Accessoires
Promotions

Promotions