missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decon™ LeLoop™ Inoculating Loops

More economical than platinum or disposable plastic loops
$278.69 - $828.06
Specifications
| Length (English) | 3 in. |
|---|---|
| Length (Metric) | 7.62 cm |
| Material | Nichrome |
| Sterility | Non-sterile |
| Product Type | Inoculating Loop |
| Catalog Number | Mfr. No. | Volume | Calibrated | Diameter (Metric) Inside | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume | Calibrated | Diameter (Metric) Inside | Price | Quantity & Availability | |||
22032088
![]() |
Decon™
MP19025 |
10 μL | Yes | 0.4 cm |
Pack of 5 Each for $828.06
Case of 5 Pack for $4,140.30
|
|
|||
22032089
![]() |
Decon™
MP19525 |
1 μL | Yes | 0.14 cm |
Pack of 5 Each for $802.28
Case of 5 Pack for $4,011.40
|
|
|||
22032090
![]() |
Decon™
MP19725 |
2.45 μL | No | 0.22 cm |
Pack of 25 Each for $278.69
|
|
|||
22032092
![]() |
Decon™
MP19925 |
3.41 μL | No | 0.29 cm |
Pack of 25 Each for $278.69
|
|
|||
22032091
![]() |
Decon™
MP19825 |
5.45 μL | No | 0.35 cm |
Pack of 25 Each for $278.69
|
|
|||
Description
More economical than platinum or disposable plastic loops
Reusable loops are made of high-quality twisted nichrome wire.
- Loops slide smoothly across agar surfaces without tearing or gouging
- The nichrome wire cools rapidly, saving time
- Loops are available calibrated or noncalibrated
- Calibrated in accordance with ASM/APHA via Evans Blue Test and include calibration certificates
- LeLoop™ loops come without holders, which are available separately
Specifications
| 3 in. | |
| Nichrome | |
| Inoculating Loop |
| 7.62 cm | |
| Non-sterile |