missing translation for 'onlineSavingsMsg'
Learn More

AlphaProTech Critical Cover™ GenPro™ Shoe Covers: Serged Seam

Catalog No. 19140200
Change view
Click to view available options
Size:
Universal
X-Large
Thread Color:
Black
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Thread Color
19140200 Universal White
191304271 X-Large Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19140200 Supplier AlphaProTech Supplier No. SH 71222 B
Only null left

Non-conductive polypropylene shoe covers are for use in non-hazardous environments. Ideal for general purpose or for facility visitors.

  • Moderate barrier to fluids, particulates and contamination
  • Available in anti-skid
  • Serged at the seam
  • Blue

Specifications

Size Universal
Material Polypropylene
Latex Free No
Color Blue
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top
Sole Type Anti-skid
Seam Type Serged
Thread Color White
For Use With (Application) Non-hazardous Environments, Facility Visitors
Packaging Quantity 150 Pairs/Cs.
Length (English) 15.75 in.
Length (Metric) 40 cm
Height (English) 7 in.
Height (Metric) 17.78 cm
Resistance Dry Particulates, Fluid
Product Type Controlled Environments Shoe Covers
Show More Show Less