Learn More
Corning™ Costar™ Snap Cap Microcentrifuge Tubes
Features a snap cap with a positive seal that allows for repeated opening and closing with flat-cap surfaces for convenient labeling. Corning™ Costar™ Snap Cap Microcentrifuge Tubes are compatible with leading microcentrifuge rotors.
Supplier: Corning™ C3621
Description
- Supplied nonsterile and are autoclavable.
- External graduations and frosted writing spot for easy sample identification.
- Positive seal design allows repeated opening and closing.
- Flat cap surface for convenient labeling.
- Withstands a maximum RCF of 17,000 x g.
- RNase-/DNase-free.
Chemical Identifiers
| Snap Cap | |
| Polypropylene | |
| Polypropylene | |
| Yes | |
| Conical | |
| Microcentrifuge Tube |
| RNase, DNase-free | |
| Autoclavable | |
| Non-sterile | |
| Snap Cap | |
| 17,000 x g |
Specifications
| Snap Cap | |
| Natural | |
| Autoclavable | |
| Natural | |
| 500/Pk. | |
| Non-sterile | |
| Snap Cap | |
| 17,000 x g |
| RNase, DNase-free | |
| Polypropylene | |
| 1.7 mL | |
| Polypropylene | |
| 5000/Pk. | |
| Yes | |
| Conical | |
| Microcentrifuge Tube |