missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Ultrapure Laminin, Mouse
GREENER_CHOICE

Numéro de catalogue. CB40239
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
CB40239 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CB40239 Fournisseur Corning™ Code fournisseur 354239
Il en reste null

Promotes cell adhesion, migration, growth, and differentiation, including neurite outgrowth

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Laminin is a major component of basement membranes. It has numerous biological activities ­including promotion of cell adhesion, migration, chemotaxis, growth, and differentiation, including neurite outgrowth.

Spécifications

Concentration 1 to 10μg/ cm2
For Use With (Application) Cell Adhesion, Cell Growth and Differentiation, Cell Migration, Neurite Outgrowth
Formulation Frozen in 0.05M Tris HCl, 0.15M NaCl pH 7.4
Molecular Weight (g/mol) 900kD (unreduced)
Name Mouse Ultrapure Laminin
pH Range 7.4
Quality Control Testing >90% by SDS-PAGE
Quantity 1 mg
Source Engelbreth Holm Swarm (EHS) Mouse tumor
Storage Requirements -70°C
Cross Reactivity Mouse
Form Liquid
Afficher plus de résultats Afficher moins de résultats