Learn More
Corning™ CellBIND™ Surface Microplates

Treated with CellBIND™ to help cells adapt to reduced serum or serum-free media and improve cell attachment for increased cell growth and yield. Corning™ CellBIND™ Surface Microplates are constructed of clear polystyrene for optimum visibility and are available in multiple well formats.
$680.14 - $1,472.54
Specifications
| Closure Included | Yes |
|---|---|
| Material | Polystyrene |
| Sterility | Sterile |
| Surface Treatment | CellBIND |
| No. per Case | 50/Cs. |
| Catalog Number | Mfr. No. | Culture Area | Capacity (Metric) | Volume (Metric) Working | Well Count | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Culture Area | Capacity (Metric) | Volume (Metric) Working | Well Count | Price | Quantity & Availability | ||
08757215
![]() |
Corning™
3336 |
3.8 cm2 | 6.9 mL | 0.76 to 1.14 mL | 12 |
Case of 50 Each for $1,137.32
|
|
||
08757214
![]() |
Corning™
3335 |
9.5 cm2 | 2.9 mL (Well) | 1.9 to 2.9 mL | 6 |
Case of 50 Each for $680.14
|
|
||
08757216
![]() |
Corning™
3337 |
1.9 cm2 | 0.57 mL (Well) | 0.38 to 0.57 mL | 24 |
Case of 50 Each for $1,298.68
|
|
||
13700435
![]() |
Corning™
3338 |
0.95 cm2 | 0.28 mL (Well) | 0.19 to 0.28 mL | 48 |
Case of 50 Each for $1,472.54
|
|
||
Description
Treated with CellBIND™ to help cells adapt to reduced serum or serum-free media and improve cell attachment for increased cell growth and yield. Corning™ CellBIND™ Surface Microplates are constructed of clear polystyrene for optimum visibility and are available in multiple well formats.
- Individual alphanumerical codes for well identification, flat bottoms
- Treated for optimal cell attachment (except where noted)
- Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
- Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
- Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
- Sterile
- Nonpyrogenic
Certifications
Certified nonpyrogenic
Specifications
| Yes | |
| Sterile | |
| 50/Cs. |
| Polystyrene | |
| CellBIND | |
| 5/Pk. |