Learn More
Corning™ CellBIND™ Surface Microplates

Treated with CellBIND™ to help cells adapt to reduced serum or serum-free media and improve cell attachment for increased cell growth and yield. Corning™ CellBIND™ Surface Microplates are constructed of clear polystyrene for optimum visibility and are available in multiple well formats.
Supplier: Corning™ 3338
Description
Treated with CellBIND™ to help cells adapt to reduced serum or serum-free media and improve cell attachment for increased cell growth and yield. Corning™ CellBIND™ Surface Microplates are constructed of clear polystyrene for optimum visibility and are available in multiple well formats.
- Individual alphanumerical codes for well identification, flat bottoms
- Treated for optimal cell attachment (except where noted)
- Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
- Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
- Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
- Sterile
- Nonpyrogenic
Certifications
Certified nonpyrogenic
Identifiants chimiques
| Yes | |
| Sterile | |
| 50/Cs. |
| Polystyrene | |
| CellBIND | |
| 5/Pk. |
Spécifications
| Clear | |
| Polystyrene | |
| 0.28 mL (Well) | |
| 0.95 cm2 | |
| Rectangular | |
| CellBIND | |
| 0.19 to 0.28 mL | |
| Lid | |
| 5/Pk. |
| Yes | |
| Flat | |
| Clear | |
| Polystyrene | |
| Sterile | |
| Multi-well Cell Culture Plate | |
| 48 | |
| 50/Cs. |