Learn More
Corning™ CellBIND™ Surface Microplates
GREENER_CHOICE
Treated with CellBIND™ to help cells adapt to reduced serum or serum-free media and improve cell attachment for increased cell growth and yield. Corning™ CellBIND™ Surface Microplates are constructed of clear polystyrene for optimum visibility and are available in multiple well formats.
Supplier:  Corning™ 3337
 View more versions of this product
Catalog No. 08757216
Only null left
Description
Description
Treated with CellBIND™ to help cells adapt to reduced serum or serum-free media and improve cell attachment for increased cell growth and yield. Corning™ CellBIND™ Surface Microplates are constructed of clear polystyrene for optimum visibility and are available in multiple well formats.
  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic
Certifications
Certified nonpyrogenic
Specifications
Chemical Identifiers
Yes
Sterile
50/Cs.
Polystyrene
CellBIND
5/Pk.
Specifications
Clear
Polystyrene
0.57 mL (Well)
1.9 cm2
Rectangular
CellBIND
0.38 to 0.57 mL
Lid
5/Pk.
Yes
Flat
Clear
Polystyrene
Sterile
Multi-well Cell Culture Plate
24
50/Cs.
Product Suggestions
Product Suggestions