missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CellBIND™ Surface Microplates
GREENER_CHOICE

Catalog No. 08757215
Change view
Click to view available options
Culture Area:
0.95 cm2
1.9 cm2
3.8 cm2
9.5 cm2
Capacity (Metric):
0.28 mL (Well)
0.57 mL (Well)
2.9 mL (Well)
6.9 mL
Volume (Metric) Working:
0.19 to 0.28 mL
0.38 to 0.57 mL
0.76 to 1.14 mL
1.9 to 2.9 mL
Well Count:
12
24
48
6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Culture Area Capacity (Metric) Volume (Metric) Working Well Count
08757215 3.8 cm2 6.9 mL 0.76 to 1.14 mL 12
08757214 9.5 cm2 2.9 mL (Well) 1.9 to 2.9 mL 6
08757216 1.9 cm2 0.57 mL (Well) 0.38 to 0.57 mL 24
13700435 0.95 cm2 0.28 mL (Well) 0.19 to 0.28 mL 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 08757215 Supplier Corning™ Supplier No. 3336
Only null left

Treated with CellBIND™ to help cells adapt to reduced serum or serum-free media and improve cell attachment for increased cell growth and yield. Corning™ CellBIND™ Surface Microplates are constructed of clear polystyrene for optimum visibility and are available in multiple well formats.

  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic

Certifications

Certified nonpyrogenic

Specifications

Closure Included Yes
Capacity (Metric) 6.9 mL
Culture Area 3.8 cm2
Material Polystyrene
Shape Round
Sterility Sterile
Surface Treatment CellBIND
Type Cell Culture Plate
Volume (Metric) Working 0.76 to 1.14 mL
Well Count 12
No. per Case 50/Cs.
No. per Pack 5/Pk.
Show More Show Less