missing translation for 'onlineSavingsMsg'
Learn More

Copper sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.995% (metals basis)

Catalog No. AA43039KS
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43039KS 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA43039KS Supplier Thermo Scientific Chemicals Supplier No. 043039KS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-50-8 | Cu | 63.55 g/mol

Sputtering targets commonly used to coat electronic components and electronic instruments. Due to its high electrical conductivity, copper is becoming increasingly popular for use in large-format, high-resolution TFT-LCD television sets. Copper is used as the electrode layer in thin-film transistors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Quantity 1 Ea.
Odor Odorless
Solubility Information Insoluble in water.
Percent Purity 99.995%
Assay (metals basis)
Chemical Name or Material Copper sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
EINECSNumber 231-159-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only