missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Nickel wire, 0.075mm (0.003in) dia
Description
Resistance alloy, negative lead for type J thermocouple
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Copper Nickel wire |
| Name Note | 0.075mm (0.003 in.) dia. |
| Avg. Mol. Wt. or Mol. Wt. Range | Cu:Ni 55:45 wt% |
| Quantity | 500 m |
| Weight | ≈0.04g/m |
| Form | Wire |
| MDL Number | MFCD00801098 |
| Synonym | Constantan |
RUO – Research Use Only