missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) carbonate basic, pure, 54-56% Cu

Numéro de catalogue. p-46571
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC196670250 25 g Glass bottle
AC196675000 500 g Glass bottle
AC196670025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC196670250 Fournisseur Thermo Scientific Chemicals Code fournisseur 196670250
Il en reste null

CAS: 12069-69-1 | CCuO3·CuO2H2 | 221.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Identifiants chimiques

CAS 12069-69-1
Molecular Formula CCuO3·CuO2H2
Molecular Weight (g/mol) 221.1
MDL Number MFCD00010976
Synonym Copper(II)hydroxide carbonate

Spécifications

Melting Point 200.0°C
Color Green
Physical Form Fine Crystalline Powder
Quantity 25 g
Linear Formula CuCO3·Cu(OH)2
Fieser 01,163
Merck Index 15, 2621
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids, ammonia, practically insoluble in alcohol
Formula Weight 221.1
Percent Purity 54 to 56% Cu
Grade Pure
Packaging Glass bottle
Density 4
Chemical Name or Material Copper(II) carbonate basic
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position c
EINECSNumber 235-113-6
RTECSNumber GL6910000
TSCA TSCA

RUO – Research Use Only