missing translation for 'onlineSavingsMsg'
Learn More

Copper ICP Standard, 1000 ppm Cu in 3% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company PCU1KN500

 View more versions of this product

Catalog No. PCU1KN-500


Only null left

Description

Description

Specifications

Specifications

Copper ICP Standard
7440-50-8 , 7697-37-2 , 7732-18-5
93.0,4.46,0.09
1 mL = 1 mg Cu
Cu
Blue
Approximately 0°C
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
500 mL
CHEBI:30052
Natural Poly Bottle
1,000ppm Cu
97.0,4.64,0.1
Liquid
MFCD00010965
< 2
Standard
[Cu]
63.55
23978

Chemical Identifiers

7440-50-8
63.55
RYGMFSIKBFXOCR-UHFFFAOYSA-N
CHEBI:30052
[Cu]
Cu
MFCD00010965
23978
copper
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions