missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper ICP Standard, 1000 ppm Cu in 3% HNO3, Ricca Chemical
$275.97 - $628.07
Chemical Identifiers
| CAS | 7440-50-8 |
|---|---|
| Molecular Formula | Cu |
| Molecular Weight (g/mol) | 63.55 |
| MDL Number | MFCD00010965 |
| InChI Key | RYGMFSIKBFXOCR-UHFFFAOYSA-N |
| PubChem CID | 23978 |
| ChEBI | CHEBI:30052 |
| IUPAC Name | copper |
| SMILES | [Cu] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PCU1KN-100
|
Ricca Chemical Company
PCU1KN100 |
100 mL | Natural LDPE Bottle |
Each for $275.97
|
|
||||
|
PCU1KN-500
|
Ricca Chemical Company
PCU1KN500 |
500 mL | Natural Poly Bottle |
Each for $628.07
|
|
||||
Chemical Identifiers
| 7440-50-8 | |
| 63.55 | |
| RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
| CHEBI:30052 | |
| [Cu] |
| Cu | |
| MFCD00010965 | |
| 23978 | |
| copper |
Specifications
| Natural LDPE Bottle | |
| 1,000ppm Cu | |
| 97.0,4.64,0.1 | |
| Liquid | |
| MFCD00010965 | |
| < 2 | |
| Standard | |
| [Cu] | |
| 63.55 | |
| 23978 | |
| Copper ICP Standard |