Learn More
Copper AA Standard, Certified, 1000ppm ±5ppm (1mL = 1mg Cu), LabChem™
Supplier: LabChem LC134001
Specifications
| Copper AA Standard | |
| Poly Bottle | |
| 97.63,2,0.37 | |
| 1g/mL | |
| 1000 ppm ±5 ppm (1mL = 1mg Cu) | |
| UN3264 | |
| Soluble in water | |
| 232.59 |
| 1000ppm (1mL = 1mg Cu) | |
| 19004-19-4 , 7697-37-2 , 7732-18-5 | |
| Passes Test | |
| Liquid | |
| Cu(NO3)2·25H2O | |
| Blue/Green | |
| 500 mL | |
| Certified |
Chemical Identifiers
| 19004-19-4 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If in skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
Warning
Recommended Storage : Room Temperature