missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper AA Standard, Certified, 1000ppm ±5ppm (1mL = 1mg Cu), LabChem™
Specifications
Specifications
| Packaging | Poly Bottle |
| CAS Max % | 97.63, 2, 0.37 |
| Identification | Passes Test |
| Density | 1g/mL |
| Physical Form | Liquid |
| Concentration | 1000 ppm ±5 ppm (1mL = 1mg Cu) |
| Linear Formula | Cu(NO3)2·25H2O |
| UN Number | UN3264 |
| Color | Blue/Green |
| Solubility | Soluble in water |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. Harmful to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Wear protective gloves, eye protection. Wash exposed skin thoroughly after handling. Avoid release to the environment. If in skin: Wash with plenty of soap and water. If skin irritation occurs: Get medical advice/attention. Take off contaminated clothing and wash before reuse. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |