Learn More
Corning™ CoolBox™ XT Systems
Provide sample cooling or freezing without ice, electricity, or batteries.
Supplier:  Corning™ 432024
 Afficher plus de versions de ce produit
Numéro de catalogue. 07210024
Il en reste null
Description
Description
Corning CoolBox XT systems are bench top cooling workstations that provide sample cooling or freezing without ice, electricity or batteries. CoolBox systems are versatile and accommodate a variety of sample formats and temperatures. The modular design enables the use of CoolRack™ and CoolSink™ thermo-conductive sample modules to hold microfuge tubes, cryogenic vials, PCR tubes or plates, assay plates and more.
  • Consists of a base, cooling cartridge/core and a lid
  • Includes a magnetized collar and a 1° to 8°C temperature indicator for visual assurance of proper temperature
  • To use with any 30 place Corning CoolRack Tubes Module
  • The internal cooling or freezing cores provide 0.5° to 4°C for over 10 hours with lid open and over 16 hours with lid closed
  • CoolBox systems may also be used with dry ice to create a compact, portable, snap freezing workstation for bacteria, virus or proteins.
Spécifications
Identifiants chimiques
Benchtop cooling system
Spécifications
Benchtop cooling system
Cool Box
30 Place
CoolBox XT
Pink
Enables Cooling for 16 Hours, Freezing for 8 Hours
Holder and One Cooling Core
3 kg
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS