Learn More
Corning™ CoolBox™ XT Systems
Provide sample cooling or freezing without ice, electricity, or batteries.
$1,969.50 - $3,010.55
| Catalog Number | Mfr. No. | Color | Includes | Product Line | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Includes | Product Line | Price | Quantity & Availability | |||
|
07210021
|
Corning™
432021 |
Purple | Holder and One Cooling Core | CoolBox XT |
Case for $1,969.50
|
|
|||
|
07210022
|
Corning™
432022 |
Green | Holder and one cooling core | CoolBox XT |
Case for $2,020.00
|
|
|||
|
07210023
|
Corning™
432023 |
Orange | Holder and One Cooling Core | CoolBox XT |
Case for $1,969.50
|
|
|||
|
07210024
|
Corning™
432024 |
Pink | Holder and One Cooling Core | CoolBox XT |
Case for $1,969.50
|
|
|||
|
07210025
|
Corning™
432025 |
Purple | Holder and Two Cooling Cores | CoolBox 2XT |
Case for $3,010.55
|
|
|||
|
07210026
|
Corning™
432026 |
Green | Holder and Two Cooling Cores | CoolBox 2XT |
Case for $2,779.82
|
|
|||
|
07210027
|
Corning™
432027 |
Orange | Holder and Two Cooling Cores | CoolBox 2XT |
Case for $3,010.55
|
|
|||
|
07210028
|
Corning™
432028 |
Pink | Holder and Two Cooling Cores | CoolBox 2XT |
Case for $3,010.55
|
|
|||
Description
Corning CoolBox XT systems are bench top cooling workstations that provide sample cooling or freezing without ice, electricity or batteries. CoolBox systems are versatile and accommodate a variety of sample formats and temperatures. The modular design enables the use of CoolRack™ and CoolSink™ thermo-conductive sample modules to hold microfuge tubes, cryogenic vials, PCR tubes or plates, assay plates and more.
- Consists of a base, cooling cartridge/core and a lid
- Includes a magnetized collar and a 1° to 8°C temperature indicator for visual assurance of proper temperature
- To use with any 30 place Corning CoolRack Tubes Module
- The internal cooling or freezing cores provide 0.5° to 4°C for over 10 hours with lid open and over 16 hours with lid closed
- CoolBox systems may also be used with dry ice to create a compact, portable, snap freezing workstation for bacteria, virus or proteins.
Specifications
| Benchtop cooling system |