missing translation for 'onlineSavingsMsg'
Learn More

Corning™ XT Starter Ice-free Coolers

Bench top cooling systems that provide sample cooling or freezing without ice, electricity, or batteries.

$220.75 - $776.81

Spécifications

Type Ice-free Cooler
Color Purple
For Use With (Application) Sample cooling or freezing without ice, electricity, or batteries
Quantity 1
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Includes Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Includes Prix Quantity & Availability  
07210014
Accessoires Afficher les documents Détails de la promotion
Corning™
432014
XT Holder, XT Cooling Core
caisse for $776.81
Il en reste null
 
07210015
Afficher les documents Détails de la promotion
Corning™
432015
XT Starter Holder Only
caisse for $220.75
Il en reste null
 
Description

Description

  • Open-platform cooler that accommodates most Corning CoolRack™ and Corning CoolSink™ modules
  • Low profile and small footprint make it ideal for use in the hood
  • Keeps samples cold (0.5°C to 4.0°C) for up to four hours
  • 1°C to 8°C temperature indicator provides visual assurance of proper temperature.
  • System may also be used with dry ice to create a compact, portable, snap-freezing workstation for bacteria, viruses, or proteins
Spécifications

Spécifications

Ice-free Cooler
Purple
Sample cooling or freezing without ice, electricity, or batteries
1
Videos
FDS
Documents

Documents