missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Controlled Environment Lab Coat

Catalog No. 19165009C
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
5X-Large
7X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19165009C Large
19165009A Small
19165009B Medium
19165009D X-Large
19165009E 2X-Large
19165009F 3X-Large
19165009G 5X-Large
19165009H 7X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19165009C Supplier Fisherbrand™ Supplier No. LC626213FS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fisherbrand microporous composite lab coat is breathable and lightweight offering comfort and protection for the wearer all day.

  • Microporous composite material
  • Breathable and lightweight
  • Non-confining tapered collar
  • Five snap closure to provide full coverage
  • Elastic wrist
  • Three pockets

Specifications

Product Type Controlled Environments Lab Coat
Size Large
Gender Unisex
Material Microporous Composite Fabric
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
Resistance Particulates
No. of Pockets 3
Collar Type Standard/Tapered
Latex Free Yes
Seam Type Serged
Sleeve Style Set-in Sleeve
For Use With (Application) Controlled and Critical Environments
Packaging Type Double Bagged
Autoclavable Not Autoclavable
Cleanroom Class Class 100 (ISO 5)
Show More Show Less