missing translation for 'onlineSavingsMsg'
Learn More

CPC™ MPC Series Sanitary Coupling Body

Catalog No. 05113156
Encompass
Change view
Click to view available options
Material:
Polysulfone
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
05113156 Polysulfone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05113156 Supplier CPC™ Supplier No. MPCK17006T39

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Color Amber
Material Polysulfone
Type Coupling Insert
Pressure Up to 60 psi, 4.1 bar
Sterilization Method Gamma sterilized
Temperature (English) Max. 300°F
Temperature (English) Min -104°F
Temperature (Metric) Max 149°C