missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobamamide, 96%, Spectrum™ Chemical
$3593.18 - $172672.32
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
18609875
|
Spectrum Chemical Mfg Cor
CO1041KGBL |
1 kg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18609876
|
Spectrum Chemical Mfg Cor
CO104100GMBL |
100 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18609878
|
Spectrum Chemical Mfg Cor
CO1045GM |
5 g |
Each for $3,593.18
|
|
|||||
Specifications
| 96% | |
| 13870-90-1 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.