missing translation for 'onlineSavingsMsg'
Learn More

Cobamamide, 96%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor CO104100GMBL

 View more versions of this product

Catalog No. 18609876

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

96%
13870-90-1
100 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.