missing translation for 'onlineSavingsMsg'
Learn More

Cobaltous Chloride, For Nitrogen (Ammonia), Nitrogen (Organic), LabChem™

Catalog No. LC132001
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC132001 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC132001 Supplier LabChem Supplier No. LC132001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7791-13-1 , 7732-18-5 , 7647-01-0
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
Synonym Cobalt (II) Chloride, Cobalt muriate hexahydrate
PubChem CID 24643 
ChEBI CHEBI:53503
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Color Pink
CAS Max % 1.2, 94.46, 4.34
Linear Formula CoCl2·6H2O
UN Number UN1789
Solubility Information Soluble in water
Formula Weight 237.92
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Hydrogen chloride; Thermal decomposition generates: Corrosive vapors; Cobalt and compounds
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Cobaltous Chloride, For Nitrogen (Ammonia), Nitrogen (Organic)
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin: Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature