missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobaltous Chloride CS, USP/EP Red Primary Solution, Ricca Chemical
1 mL = 59.5 mg CoCl2.6H2O
$256.36 - $645.79
Chemical Identifiers
| CAS | 7791-13-1 |
|---|---|
| Molecular Formula | Cl2CoH12O6 |
| Molecular Weight (g/mol) | 237.92 |
| MDL Number | MFCD00149652 |
| InChI Key | GFHNAMRJFCEERV-UHFFFAOYSA-L |
| IUPAC Name | λ2-cobalt(2+) hexahydrate dichloride |
| SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Co++] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
2210-4
|
Ricca Chemical Company
22104 |
120 mL | Amber Glass Bottle |
Each for $256.36
|
|
||||
|
2210-16
|
Ricca Chemical Company
221016 |
500 mL | Amber Glass Bottle |
Each for $398.23
|
|
||||
|
2210-32
|
Ricca Chemical Company
221032 |
1 L | Amber Glass Bottle |
Each for $645.79
|
|
||||
Chemical Identifiers
| 7791-13-1 | |
| 237.92 | |
| GFHNAMRJFCEERV-UHFFFAOYSA-L | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Co++] |
| Cl2CoH12O6 | |
| MFCD00149652 | |
| λ2-cobalt(2+) hexahydrate dichloride |
Specifications
| Red | |
| Cl2CoH12O6 | |
| GFHNAMRJFCEERV-UHFFFAOYSA-L | |
| λ2-cobalt(2+) hexahydrate dichloride | |
| 1 mL = 59.5 mg CoCl2·6H2O | |
| Amber Glass Bottle | |
| Liquid |