missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) chromate, Thermo Scientific™

Numéro de catalogue. AA3920318
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AA3920318 50 g
AA3920309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AA3920318 Fournisseur Thermo Scientific Chemicals Code fournisseur 3920318
Il en reste null

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 13455-25-9
Molecular Formula CoCrO4
Molecular Weight (g/mol) 174.93
MDL Number MFCD00053962
InChI Key XTUHPOUJWWTMNC-UHFFFAOYSA-N
Synonym cobalt ii chromate, cobalt 2+ ; dioxido dioxo chromium, cobaltous chromate iii, cobalt chromate ous, cobaltous chromate, acmc-1c0r0, cobalt 2+ chromate 2-, chromic acid h2cro4 , cobalt 2+ salt 1:1
PubChem CID 61614
IUPAC Name cobalt(2+);dioxido(dioxo)chromium
SMILES [Co++].[O-][Cr]([O-])(=O)=O

Spécifications

Melting Point (decomposition)
Physical Form Lump
Quantity 50 g
UN Number UN3087
Solubility Information Soluble in alkali and ammomium hydroxide
Formula Weight 174.93
Odor Odorless
Chemical Name or Material Cobalt(II) chromate
Hazard Category H272-H314-H317-H334-H335-H350i
Hazard Statement GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P220-P221-P260-P264b-P271-P272-P280g-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: II
EINECSNumber 236-651-4
TSCA Yes
Recommended Storage Ambient temperatures