missing translation for 'onlineSavingsMsg'
Learn More

Cobalt Chloride, 1.2% (w/v) in 10% (v/v) Hydrochloric Acid, Ricca Chemical

Catalog No. 218016
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
218016 500 mL Natural Poly Bottle
218032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 218016 Supplier Ricca Chemical Company Supplier No. 218016

Chemical Identifiers

CAS 7791-13-1 , 7732-18-5 , 7647-01-0
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
Synonym Cobalt (II) Chloride, Cobalt muriate hexahydrate
PubChem CID 24643
ChEBI CHEBI:53503
IUPAC Name λ2-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Color Pink
CAS Min % 1.04, 87.0, 10.23
CAS Max % 1.08, 90.0, 10.65
Percent Purity 1.2%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution with 10% (v/v) Hydrochloric Acid
pH <1
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 1.2% (w/v)
Chemical Name or Material Cobalt Chloride
Show More Show Less