missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Clusterin ELISA Kit

ELISA

$910.00

Spécifications

Accession Number P10909
Assay Range 19.53 to 20,000 pg/mL
Assay Sensitivity 15 pg/mL
Calibration Range 15 pg/mL to 20000 pg/mL
Assay Solid-phase sandwich ELISA
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
EHCLU
Encompass
Afficher les documents Détails de la promotion
Invitrogen™
EHCLU
96 Tests
paquet for $910.00
Il en reste null
 
Description

Description

The Human Clusterin ELISA quantitates Hu Clusterin in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Clusterin. Principle of the method The Human Clusterin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Isoform 1 functions as extracellular chaperone that prevents aggregation of nonnative proteins. Prevents stress-induced aggregation of blood plasma proteins. Inhibits formation of amyloid fibrils by APP, APOC2, B2M, CALCA, CSN3, SNCA and aggregation-prone LYZ variants (in vitro). Does not require ATP. Maintains partially unfolded proteins in a state appropriate for subsequent refolding by other chaperones, such as HSPA8/HSC70. Does not refold proteins by itself. Binding to cell surface receptors triggers internalization of the chaperone-client complex and subsequent lysosomal or proteasomal degradation. Secreted isoform 1 protects cells against apoptosis and against cytolysis by complement. Intracellular isoforms interact with ubiquitin and SCF (SKP1-CUL1-F-box protein) E3 ubiquitin-protein ligase complexes and promote the ubiquitination and subsequent proteasomal degradation of target proteins. Promotes proteasomal degradation of COMMD1 and IKBKB. Modulates NF-kappa-B transcriptional activity. Nuclear isoforms promote apoptosis. Mitochondrial isoforms suppress BAX-dependent release of cytochrome c into the cytoplasm and inhibit apoptosis. Plays a role in the regulation of cell proliferation.
Spécifications

Spécifications

P10909
15 pg/mL
Solid-phase sandwich ELISA
ELISA
Plasma, Serum, Supernatant
Human
Colorimetric Microplate Reader
AAG4,APO-J,APOJ,CLI1,CLU2,KUB1,NA1/NA2,SGP-2,SGP2,SP-40,TRPM-2,TRPM2,CLU
<10
HRP
Neurobiology
2°C to 8°C
Apolipoprotein J
4 hrs 45 mins
19.53 to 20,000 pg/mL
15 pg/mL to 20000 pg/mL
HRP
Mitochondrial Proteins
This ELISA antibody pair detect human Clusterin. Other species not determined.
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
1191
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
Plasma 1 μL, Serum 1 μL, Supernatant 100 μL
AAG4, APO-J, APOJ, CLI1, CLU2, KUB1, NA1/NA2, SGP-2, SGP2, SP-40, TRPM-2, TRPM2, CLU
1 hrs 20 mins
Videos
FDS
Documents

Documents