missing translation for 'onlineSavingsMsg'
Learn More

Clomiphene citrate salt, MilliporeSigma™ Supelco™

Clomiphene is an semisynthetic antiestrogen, widely used in the treatment of infertility by inducing ovulation and stimulating sperm production. It is also used in the treatment of breast cancer.

$181.59 - $444.96

Specifications

CAS 50-41-9
Physical Form Neat
Linear Formula C26H28 ClNO · C6H8O7
MDL Number MFCD00058322
Synonym 2-(4-[2-Chloro-1,2-diphenylethenyl]phenoxy)-N,N-diethylethanamine
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111012207
View Documents
Merck Emd Millipore
C62721G
1 g
Each for $181.59
Only null left
 
111012208
View Documents
Merck Emd Millipore
C627210G
10 g
Each for $444.96
Only null left
 
Description

Description

Pharmaceutical (small molecule)
Specifications

Specifications

50-41-9
C26H28 ClNO · C6H8O7
2-(4-[2-Chloro-1,2-diphenylethenyl]phenoxy)-N,N-diethylethanamine
598.08
Neat
MFCD00058322
Analytical Standard
Analytical Standard
Videos
SDS
Documents

Documents