CAS RN 50-41-9
CAS RN 50-41-9
IUPAC Name:
(2-{4-[(1Z)-2-chloro-1,2-diphenylethenyl]phenoxy}ethyl)diethylamine; 2-hydroxypropane-1,2,3-tricarboxylic acid
Synonyms:
clomid
Molecular Weight (g/mol):
598.09
Molecular Formula:
C32H36ClNO8
InChi Key:
PYTMYKVIJXPNBD-OQKDUQJOSA-N
SMILES:
OC(=O)CC(O)(CC(O)=O)C(O)=O.CCN(CC)CCOC1=CC=C(C=C1)C(=C(/Cl)C1=CC=CC=C1)\C1=CC=CC=C1
1
–
1
of
1
results
1
Clomiphene citrate
CAS: 50-41-9 Molecular Formula: C32H36ClNO8 Molecular Weight (g/mol): 598.089 MDL Number: MFCD00058322 InChI Key: PYTMYKVIJXPNBD-BTKVJIOYSA-N Synonym: clomifene citrate,clomiphene citrate,clomid,cis-clomiphene citrate,clomiphene a citrate,zuclomid,chloramiphene,milophene,serophene,z-clomiphene citrate PubChem CID: 6420009 IUPAC Name: 2-[4-[(E)-2-chloro-1,2-diphenylethenyl]phenoxy]-N,N-diethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: CCN(CC)CCOC1=CC=C(C=C1)C(=C(C2=CC=CC=C2)Cl)C3=CC=CC=C3.C(C(=O)O)C(CC(=O)O)(C(=O)O)O