missing translation for 'onlineSavingsMsg'
Learn More

Clomiphene citrate salt, MilliporeSigma™ Supelco™

Catalog No. 111012208
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012208 10 g
111012207 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012208 Supplier Merck Emd Millipore Supplier No. C627210G
Only null left

Clomiphene is an semisynthetic antiestrogen, widely used in the treatment of infertility by inducing ovulation and stimulating sperm production. It is also used in the treatment of breast cancer.

Pharmaceutical (small molecule)

Specifications

CAS 50-41-9
Physical Form Neat
Linear Formula C26H28 ClNO · C6H8O7
MDL Number MFCD00058322
Synonym 2-(4-[2-Chloro-1,2-diphenylethenyl]phenoxy)-N,N-diethylethanamine
Type Analytical Standard
Quantity 10 g
Formula Weight 598.08
Grade Analytical Standard