missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clomiphene citrate salt, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 50-41-9 |
| Physical Form | Neat |
| Linear Formula | C26H28 ClNO · C6H8O7 |
| MDL Number | MFCD00058322 |
| Synonym | 2-(4-[2-Chloro-1,2-diphenylethenyl]phenoxy)-N,N-diethylethanamine |
| Type | Analytical Standard |
| Quantity | 1 g |
| Formula Weight | 598.08 |
| Grade | Analytical Standard |