missing translation for 'onlineSavingsMsg'
Learn More

Carson™ Clip and Flip™ Clip-On, Flip-Up Magnifying Lenses

Wearable, hands-free magnifiers that are easy to use and suitable for any outdoorsman or person with low vision needs.

Supplier:  Carson™ OD12

Encompass

 Afficher plus de versions de ce produit

Numéro de catalogue. S98159



Il en reste null

Description

Description

  • Wearable, hands-free magnifiers that are easy to use and suitable for any outdoorsman
  • Sturdy, durable clip allows any pair of glasses to be transformed into magnifying lenses
  • The clip's flexibility also allows the user to easily switch from the magnifier to regular glasses with one quick flip
  • At 0.6 ounces, these lenses put no excess weight or strain on the user
  • Includes a protective pouch for safe and easy transportation
Spécifications

Identifiants chimiques

2 in.
Acrylic
Protective Pouch
4.7 x 2.4 x 1.5 in.
Lens, Magnifying

Spécifications

Acrylic
4.7 x 2.4 x 1.5 in.
Protective Pouch
1.75x
Lens, Magnifying
2 in.
+3.00
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions