missing translation for 'onlineSavingsMsg'
Learn More

Clindamycin Hydrochloride USP, Medisca™
GREENER_CHOICE

Catalog No. 18-048-078
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
1 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18048078 5 g
18048059 100 g
18048060 500 g
18048079 25 g
18048080 1 kg
18048104 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18048078 Supplier Medisca Pharmaceutique Inc Supplier No. 000503
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Clindamycin Hydrochloride USP is a white or practically white, crystalline powder. It is odorless to having a faint mercaptan-like odor. Stable in the presence of air and light.

Chemical Identifiers

CAS 21462-39-5

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Crystalline Powder
Color White
Quantity 5 g
Percent Purity ≥ 800 μg
Grade USP