missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Clear Vinyl Bib Apron

Catalog No. S24922
Change view
Click to view available options
Size:
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
S24922 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24922 Supplier Eisco™ Supplier No. VNAPCH

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Clear vinyl bib apron protects clothes during laboratory exercises. Ideal for use in labs, kitchens, or shops.

  • Basic transparent vinyl apron
  • Perfect for biology, chemistry, and general science laboratories
  • Protects clothes from liquid spills and splashes
  • Acid resistant

Specifications

Type Bib Apron
Size Medium
Material Vinyl
Color Clear
Disposable Reusable
Closure Type Hook and Loop (Rear)
Sterility Non-sterile
Thickness 6.7 mil (0.17 mm)
For Use With (Application) Labs, kitchens or shops
Seam Type Seamless