missing translation for 'onlineSavingsMsg'
Learn More

Purus™ Pure Comfort Cleanroom Socks
GREENER_CHOICE

Catalog No. 17456039
Change view
Click to view available options
Disposable:
Reusable
Single-use
Size:
One Size
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Disposable Size
17456039 Single-use One Size
19125961 Reusable Small
19125962 Reusable Medium
19125963 Reusable Large
19125964 Reusable X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17456039 Supplier Purus™ Supplier No. CRSD

Purus™ Cleanroom Socks are designed for cleanroom use and comfort.

  • Disposable, single-use sock
  • Made from 100% polyester
  • Cleanroom compatible
  • Comfortable for extended use
  • No particle generation

Specifications

Color White
For Use With (Application) Pharmaceutical, Biotechnology, Cleanrooms, Hospital, Semiconductors
Material Polyester
Size One Size
Disposable Single-use
Sterility Non-sterile
Packaging Quantity 400 Pair/Cs
Product Type Cleanroom Socks
Cleanroom Class Class 10-1000 (ISO 4-6)