missing translation for 'onlineSavingsMsg'
Learn More

PURUS™ Cleanroom Shoes

Catalog No. 19125959
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19125959 2X-Large
19125954 X-Small
19125955 Small
19125956 Medium
19125957 Large
19125958 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19125959 Supplier PURUS™ Supplier No. CRSHWXXL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PURUS™ Cleanroom Shoes provide comfort and durability when working in cleanroom environments.

  • Smooth surface for day-long comfort
  • Antistatic
  • Autoclavable
  • Suitable for use in ISO 4 to 6 (Class 10 to Class 1000) cleanroom
  • Non-slip sole
  • Special conical design prevents spilled liquids from entering clog
  • Heel strap
  • Anatomic Heel Basin
  • Toe Grabber

Specifications

Certifications/Compliance ASTM F2913
Color White
For Use With (Application) Pharmaceutical, Biotech, Semiconductor, Cleanrooms, Hospitals
Material Thermoplastic Elastomer
Size 2X-Large
Toe Style Plain Toe
Packaging Quantity 6/Cs.
Product Type Cleanroom Shoes