Learn More
PURUS™ Clean Image Cleanroom Notebooks
GREENER_CHOICE
CleanImage Notebooks are ideal for writing and data recording in cleanroom environments.
Supplier:  PURUS™ PNB CI 5.5X8.5 2220 L
 Afficher plus de versions de ce produit
Numéro de catalogue. 19125848
Il en reste null
Description
Description
For writing in cleanroom environments
  • Cleanroom processed and packaged
  • Suitable for use in ISO 4 to 6 (Class 10 to Class 1000) cleanrooms
  • Propriatary Binding Technology without organic fillers
  • Recyclable
  • College Rule
  • Amide and Silicone Free
Order Info
Available in 3 sizes
Compliance
Suitable for use in ISO 4 to 6 (Class 10 to Class 1000)
Spécifications
Identifiants chimiques
Yes
White
Clear
Page weight is 22 lb./105 gsm
Spécifications
Synthetic Latex Substrate
White
Purus™ Cleanroom Notebook
For writing in cleanroom environments
Page weight is 22 lb./105 gsm
Yes
Clear
5.5 x 8.5 in.
20
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS