missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ ArcGuard™ Class 00 Rubber Voltage Gloves Premium Kit

Catalog No. 170011998
Change view
Click to view available options
Size:
10
11
8
9
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170011998 11
170011995 8
170011996 9
170011997 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 170011998 Supplier National Safety Apparel™ Supplier No. KITGC0011AG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Premium kit includes a pair of electrical insulating rubber gloves, fire-resistant knit gloves, leather glove protectors, and a glove bag. Class 00 insulating gloves will protect workers from up to 500 V.

  • Electrical insulating gloves protect against electrical shock and other arc flash hazards
  • Wear with included leather glove protectors for additional resistance against mechanical hazards
  • FR knit gloves stretch for dexterity and can be worn for another layer of protection
  • Compliant with ANSI/ASTM D120 and NFPA 70E standards
  • Class 00 gloves offer protection from up to 500 V
  • Gloves should be used within one year of marked test date
  • When not in use, lay flat in the included protective bag
  • Store away from direct sunlight, high heat, and humidity

Specifications

Certifications/Compliance ANSI/ASTM D120, Class 00, Type I, NFPA 70E, ASTM F1506, OSHA 1910.269
Applications Industrial
Color Black
Material Rubber, Leather
Product Type Electrical Insulating Gloves Kit
Size 11
For Use With (Application) Electrical Utility Work, Shock Protection
Glove Cuff Style Rolled
Includes Electrical Insulating Rubber Gloves, Fire-Resistant Knit Gloves, Leather Glove Protectors, and a Glove Bag
Series KITGC00BAG
Voltage 500 V
Show More Show Less