missing translation for 'onlineSavingsMsg'
Learn More

Chlorocyclohexane, 98%

Catalog No. AAL0434930
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0434930 250 g
AAL043490B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0434930 Supplier Thermo Scientific Chemicals Supplier No. L0434930
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 542-18-7 | C6H11Cl | 118.604 g/mol

Chlorocyclohexane is used as the raw material in synthesis agrochemical and medicine chemical engineering products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorocyclohexane is used as the raw material in synthesis agrochemical and medicine chemical engineering products.

Solubility
Soluble in water.0.02 g/L (20°C). Soluble in alcohol and diethyl ether.

Notes
Incompatible materials are Strong oxidizing agents, Strong bases.

Chemical Identifiers

CAS 542-18-7
Molecular Formula C6H11Cl
Molecular Weight (g/mol) 118.604
MDL Number MFCD00003822
InChI Key UNFUYWDGSFDHCW-UHFFFAOYSA-N
Synonym cyclohexyl chloride, cyclohexane, chloro, monochlorocyclohexane, chlorcyclohexan, chlorzyklohexan, unii-pnv8c821fh, pnv8c821fh, chlor-cyclohexane, cyclohexylchloride, chloro-cyclohexane
PubChem CID 10952
ChEBI CHEBI:39156
IUPAC Name chlorocyclohexane
SMILES C1CCC(CC1)Cl

Specifications

Melting Point -44°C
Density 1
Boiling Point 141°C to 143°C
Flash Point 28°C (82°F)
Odor Unpleasant
Refractive Index 1.462
Quantity 250 g
UN Number UN1993
Beilstein 1900796
Merck Index 14,2732
Solubility Information Soluble in water.0.02g/L (20°C). Soluble in alcohol and diethyl ether.
Formula Weight 118.61
Percent Purity 98%
Chemical Name or Material Chlorocyclohexane
Show More Show Less
Hazard Category H226-H315-H319-H332-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 208-806-6
RTECSNumber GU8685000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only