missing translation for 'onlineSavingsMsg'
Learn More

Chlorine Buffer Reagent 2, Reagents

Numéro de catalogue. 01335708
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
01335708 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 01335708 Fournisseur Labchem Inc Code fournisseur CC0043001A
Il en reste null

Chlorine Buffer Reagent 2, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Spécifications

Chemical Name or Material Chlorine Buffer Reagent 2
Color Colorless
CAS 7732-18-5, 7778-77-0, 7558-79-4
Boiling Point 100°C
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0799
Quantity 1 L
Sterility Non-sterile
Physical Form Liquid