missing translation for 'onlineSavingsMsg'
Learn More

Chloride Standard, 5 ppm Cl-, Ricca Chemical

$58.27 - $348.19

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
R1941500500
View Documents
Ricca Chemical Company
R1941500500A
500 mL Natural Poly Bottle
Each for $348.19
Only null left
 
R19415001A
View Documents
Ricca Chemical Company
R19415001A
1 L Natural Poly Bottle
Each for $58.27
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
CHEBI:26710
[Na+].[Cl-]
ClNa
MFCD00003477
5234
sodium chloride

Specifications

Natural Poly Bottle
100,0.0008
1
ClNa
Colorless
0°C
Miscible with water
[Na+].[Cl-]
58.44
5234
Chloride Standard, 5 ppm Cl-
7647-14-5,7732-18-5
100,0.0008
Liquid
MFCD00003477
6.7 to 7.3
100°C
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
500 mL
CHEBI:26710
Videos
Safety and Handling

Safety and Handling

ShelfLife : 6 months

Recommended Storage : Ambient

SDS
missing translation for 'documents'

missing translation for 'documents'