missing translation for 'onlineSavingsMsg'
Learn More

Chloride Standard, 5 ppm Cl-, Ricca Chemical

Supplier:  Ricca Chemical Company R19415001A

 View more versions of this product

Catalog No. R19415001A


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Chloride Standard
Natural Poly Bottle
Odorless
Liquid
MFCD00003477
100°C
FAPWRFPIFSIZLT-UHFFFAOYSA-M
Water
58.44
5234
1mL = 0.005mg Cl-, 5ppm Cl-
7647-14-5,7732-18-5
5.00 ± 0.05ppm
ClNa
Colorless
Infinitely soluble in water
[Na+].[Cl-]
sodium chloride
1 L
CHEBI:26710

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
CHEBI:26710
[Na+].[Cl-]
ClNa
MFCD00003477
5234
sodium chloride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions