missing translation for 'onlineSavingsMsg'
Learn More

Chloride Standard, 5 ppm Cl-, Ricca Chemical

Supplier:  Ricca Chemical Company R1941500500A

 View more versions of this product

Catalog No. R1941500500


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Chloride Standard, 5 ppm Cl-
7647-14-5,7732-18-5
100,0.0008
Liquid
MFCD00003477
6.7 to 7.3
100°C
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
500 mL
CHEBI:26710
Natural Poly Bottle
100,0.0008
1
ClNa
Colorless
0°C
Miscible with water
[Na+].[Cl-]
58.44
5234

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
CHEBI:26710
[Na+].[Cl-]
ClNa
MFCD00003477
5234
sodium chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 6 months

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions