missing translation for 'onlineSavingsMsg'
Learn More
Learn More
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
18048067
![]() |
N/A | 500 g |
Each for $1,165.40
|
|
|||||
18048085
![]() |
N/A | 25 g |
Each for $82.81
|
|
|||||
18048086
![]() |
N/A | 100 g |
Each for $245.34
|
|
|||||
18048087
![]() |
N/A | 25 kg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
18048107
![]() |
N/A | 1 kg |
Each for $2,284.81
|
|
|||||
Description
Chloramphenicol USP comes as fine, white to grayish-white or yellowish-white, needle-like crystals or elongated plates.
Chemical Identifiers
| 56-75-7 |
Specifications
| α-Carboxybenzyl Penicillin Disodium Salt | |
| 56-75-7 | |
| 97.0 to 103.0% |
| Crystals | |
| Beige or Gray/Yellow to White | |
| USP |
