missing translation for 'onlineSavingsMsg'
Learn More

Chloramphenicol USP , Medisca™
GREENER_CHOICE

Catalog No. 18048107
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
25 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18048107 1 kg
18048067 500 g
18048085 25 g
18048086 100 g
18048087 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18048107 Supplier Medisca Pharmaceutique Inc Supplier No. 020309
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chloramphenicol USP comes as fine, white to grayish-white or yellowish-white, needle-like crystals or elongated plates.

Chemical Identifiers

CAS 56-75-7

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Crystals
Color Beige or Gray/Yellow to White
Quantity 1 kg
Percent Purity 97.0 to 103.0%
Grade USP